2-Propanethiol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isopropyl mercaptan, Isopropanethiol, 2-Mercaptopropane

13144-0025 13144-0500 13144-5000
TS13144-0025EA 224 USD
TS13144-0025 TS13144-0500 TS13144-5000
2-Propanethiol 98%
2-Propanethiol
Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 57…60 °C (760 mmHg)
Melting Pt: –131 °C
Density: 0.82 g/cm³
Flash Pt: –34 °C
Storage Temperature: Ambient
MDL Number: MFCD00004863
CAS Number: 75-33-2
EINECS: 200-861-4
UN: 2402
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4270 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR