1-Octanethiol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15748-0010
15748-0020
15748-0025
15748-2500
TS15748-0010EA
63
USD
TS15748-0010
TS15748-0020
TS15748-0025
TS15748-2500
1-Octanethiol 97%
1-Octanethiol
Formula:
C₈H₁₈S Boiling Pt: 197…200 °C (760 mmHg) Melting Pt: –49 °C Density: 0.84 g/cm³ Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004912 CAS Number: 111-88-6 EINECS: 203-918-1 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4525 to 1.4545 (20°C, 589 nm) |
GC | ≥96.0 % |
Titration with NaOH | ≥96.0 to ≤104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...