Ethanethiol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl mercaptan, 1-Mercaptoethane

11786-0025 11786-0500 11786-5000
TS11786-0025EA 222 USD
TS11786-0025 TS11786-0500 TS11786-5000
Ethanethiol 99+%
Ethanethiol
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 35 °C (760 mmHg)
Melting Pt: –148 °C
Density: 0.83 g/cm³
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00004887
CAS Number: 75-08-1
EINECS: 200-837-3
UN: 2363
ADR: 3,I
Merck Index: 12,03771

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4296 to 1.4316 (20°C, 589 nm)
GC ≥99.0 %
Cloud point ≤20.0°C
Calcium (Ca) ≤30 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR