3-Pentanol 98%

Supplier: Thermo Scientific Chemicals
Warning

12999-0250 12999-1000 12999-5000
TS12999-0250EA 49 USD
TS12999-0250 TS12999-1000 TS12999-5000
3-Pentanol 98%
3-Pentanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 114…116 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.815 g/cm³
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004574
CAS Number: 584-02-1
EINECS: 209-526-7
UN: 1105
ADR: 3,III
Merck Index: 13,07197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
GC ≥97.5 %
Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR