(±)-3-Heptanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14909-0250
14909-1000
14909-5000
TS14909-0250EA
53
USD
TS14909-0250
TS14909-1000
TS14909-5000
(±)-3-Heptanol 98%
(±)-3-Heptanol
Formula:
C₇H₁₆O Boiling Pt: 156 °C (1013 hPa) Melting Pt: –70 °C Density: 0.818 g/cm³ Flash Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004586 CAS Number: 589-82-2 EINECS: 209-661-1 UN: 1987 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4220 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...