1,2,3,4-Butanetetracarboxylic acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

36676-1000 36676-5000
TS36676-1000EA 55 USD
TS36676-1000 TS36676-5000
1,2,3,4-Butanetetracarboxylic acid 99+%
1,2,3,4-Butanetetracarboxylic acid
Formula: C₈H₁₀O₈
MW: 234.16 g/mol
Melting Pt: 195…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00002722
CAS Number: 1703-58-8
EINECS: 216-938-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 188 to 192 °C
Titration with NaOH 99.0 to 101.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR