Sodium propionate 99.0-100.5%

Supplier: Thermo Scientific Chemicals

Synonyms: Propionic acid sodium salt, Sodiumpropionate

14901-0010 14901-0050 14901-0250
TS14901-0010EA 71 USD
TS14901-0010 TS14901-0050 TS14901-0250
Sodium propionate 99.0-100.5%
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 285…286 °C
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS Number: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Titration with HClO4 99.0 to 100.5 % (On dried substance)
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR