Butyric acid 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Butanoic acid

10811-0010 10811-0025 10811-0100 10811-1000
TS10811-0010EA 73 USD
TS10811-0010 TS10811-0025 TS10811-0100 TS10811-1000
Butyric acid 99+%
Butyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 162…165 °C (760 mmHg)
Melting Pt: 7…5 °C
Density: 0.96 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8,III
Merck Index: 13,01593

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3965 to 1.3995 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≤0.2 % Isobutyric acid
Color scale ≤15 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR