Isobutyl acetate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylpropyl acetate

25983-0010 25983-0025 25983-0250
TS25983-0010EA 60 USD
TS25983-0010 TS25983-0025 TS25983-0250
Isobutyl acetate 98%
Isobutyl acetate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.873 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008932
CAS Number: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3,II
Merck Index: 13,05147

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20°C, 589 nm)
Water ≤0.03 %
Density (20°C) 0.871 to 0.875 kg/l
GC ≥97.5 %
Color scale ≤10 APHA
Acidity (CH3COOH) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR