(±)-4-Vinylcyclohexene stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-4-Ethenyl-1-cyclohexene, 4-Vinyl-1-cyclohexene

14088-0050 14088-5000
TS14088-0050EA 22 USD
TS14088-0050 TS14088-5000
(±)-4-Vinylcyclohexene stabilized
(±)-4-Vinylcyclohexene
Formula: C₈H₁₂
MW: 108.18 g/mol
Boiling Pt: 126…127 °C (760mmHg)
Melting Pt: –101 °C
Density: 0.832
Flash Pt: 16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001576
CAS Number: 100-40-3
EINECS: 202-848-9
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4660 (20 °C, 589 nm)
GC ≥96.0 %
Stabilizer 25 to 200 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR