(±)-2-Pentanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Methyl propyl carbinol, (±)-2-Hydroxy pentane, DL-2-Pentanol, (±)-Propyl methyl carbinol

12998-0010 12998-0025 12998-5000
TS12998-0010EA 118 USD
TS12998-0010 TS12998-0025 TS12998-5000
(±)-2-Pentanol 98%
(±)-2-Pentanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 118…119 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.812 g/cm³
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00004579
CAS Number: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3,III
Merck Index: 12,07258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc37771666c375a7768be6819fbc64e80617a202&vid=cc73accd09f3b4e43eedad1fe603cf52c119ff25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
Water ≤0.3 %
GC ≥97.5 %
Residue after evaporation ≤0.003 %
Acidity (CH3COOH) ≤0.005 %
Ketone ≤1 % (as methyl propyl ketone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR