(±)-2-Hexanol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane

15629-0100 15629-0500 15629-2500
TS15629-0100EA 39 USD
TS15629-0100 TS15629-0500 TS15629-2500
(±)-2-Hexanol 99%
(±)-2-Hexanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004585
CAS Number: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db7ec887376d519b077785cdead15c1aed20d58&vid=5c0c76fb523ec80ddb5134157d5d19ce7a63dcc3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4125 to 1.4155 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR