Presaturated with Quaternary ammonium and ethyl alcohol.
- Laundered knitted 100% polyester fabric has extremely low levels of particles and fibres
- 1 minute disinfectant contact time is easy to maintain compliance and appropriate dwell time
- Presaturated with disinfectant reduces VOC emissions and solvent use
- Resealable pouch preserves cleanliness and solvent saturation levels
PROSAT® Wipes with CyQuanol™ are intended for Life Science cleanroom manufacturers who want to effectively maintain cGMP compliance by maximizing efficiency and minimizing inconsistency in their contamination control process.
PROSAT® Polynit Heatseal Wipes with CyQuanol™ are manufactured from 100% knitted polyester with laser sealed edges for very low levels of particles and fibres. These wipes are EPA registered with proven efficacy claims against bacteria, viruses, and fungi. With a short contact time of 1 minute, these wipes make compliance to disinfection protocols more manageable, as microbial kill claims can be achieved without reapplication. Using presaturated wipes delivers a controlled amount of disinfectant to surfaces every time compared to traditional spray methods. Presaturated wipes ensure repeatability and reproducibility of disinfectant protocols to improve process consistency.
PROSAT® Wipes with CyQuanol™ are also available with melt blown polypropylene. Both substrates are available sterile for the most critical areas.