Favipiravir

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide, T-705, 6-Fluoro-3-hydroxypyrazine-2-carboxamide

46869-1000 468695000
TS46869-1000EA 188 USD
TS46869-1000 TS468695000
Favipiravir
Favipiravir
Formula: C₅H₄FN₃O₂
MW: 157.09 g/mol
Boiling Pt: 52.6°C (760mmHg)
Storage Temperature: Ambient
MDL Number: MFCD16879084
CAS Number: 259793-96-9

Order Now

Specification Test Results

Appearance White to yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥ 96.0%
Water ≤ 2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR