Celastrol

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tripterin

46834-1000 46834-5000
TS46834-1000EA 143 USD
TS46834-1000 TS46834-5000
Celastrol
Celastrol
Formula: C₂₉H₃₈O₄
MW: 450.62 g/mol
Melting Pt: 203°C
Storage Temperature: Refrigerator
MDL Number: MFCD03424073
CAS Number: 34157-83-0
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Light yellow to amber crystalline powder
Infrared spectrum Conforms
Melting point 200°C to 205°C
HPLC ≥ 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR