Posaconazole

Supplier: Thermo Scientific Chemicals
Warning

46766-0010 467660050
TS46766-0010EA 135 USD
TS46766-0010 TS467660050
Posaconazole
Posaconazole
Formula: C₃₇H₄₂F₂N₈O₄
MW: 700.79 g/mol
Melting Pt: 170°C…179°C
Storage Temperature: Refrigerator
MDL Number: MFCD00941162
CAS Number: 171228-49-2

Order Now

Specification Test Results

Appearance White to light yellow to beige powder or crystals
Infrared spectrum Conforms
Melting point 165°C to 170°C
Water ≤ 1%
Specific optical rotation -24° to -30° (20°C, 589nm) (1% in CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR