4-[(2-Tetrahydropyranyl)oxy]phenol

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Deoxyarbutin, 4-((Tetrahydro-2H-pyran-2-yl)oxy)phenol, 4-(Tetrahydropyran-2-yloxy)phenol

46760-0050 46760-0250
TS46760-0050EA 44 USD
TS46760-0050 TS46760-0250
4-[(2-Tetrahydropyranyl)oxy]phenol
4-[(2-Tetrahydropyranyl)oxy]phenol
Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09970981
CAS Number: 53936-56-4

Order Now

Specification Test Results

Appearance White to beige powder
Infrared spectrum Conforms
Melting point 83°C to 88°C
HPLC ≥ 97.5%
Water ≤ 2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR