Cimetidine

Supplier: Thermo Scientific Chemicals

46752-0050 467520250
TS46752-0050EA 57 USD
TS46752-0050 TS467520250
Cimetidine
Cimetidine
Formula: C₁₀H₁₆N₆S
Melting Pt: 141°C…143°C
Storage Temperature: Ambient
MDL Number: MFCD00133296
CAS Number: 51481-61-9
EINECS: 257-232-2
Merck Index: 13.23

Order Now

Specification Test Results

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
Melting point 141°C to 143°C
HPLC ≥ 97.5%
Loss on drying ≤ 0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR