Metamizole sodium monohydrate

Supplier: Thermo Scientific Chemicals
Warning

46748-2500 467480010
TS46748-2500EA 117 USD
TS46748-2500 TS467480010
Metamizole sodium monohydrate
Metamizole sodium monohydrate
Formula: C₁₃H₁₆N₃NaO₄S·H₂O
MW: 351.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150293
CAS Number: 5907-38-0
Merck Index: 13.3385

Order Now

Specification Test Results

Appearance White to light yellow to light beige powder or crystals
Infrared spectrum Conforms
Melting point 211°C to 216°C
HPLC ≥ 97.5%
Water 4.5 to 5.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR