Desonide

Supplier: Thermo Scientific Chemicals

Synonyms: (11beta,16alpha)-11,21-Dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Budesonide Acetonide

46745-0010 467450050 467452500
TS46745-0010EA 67 USD
TS46745-0010 TS467450050 TS467452500
Desonide
Desonide
Formula: C₂₄H₃₂O₆
MW: 416.51 g/mol
Melting Pt: 275°C
Storage Temperature: Ambient
MDL Number: MFCD00866151
CAS Number: 638-94-8
EINECS: 211-351-6

Order Now

Specification Test Results

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Loss on drying ≤ 0.5% (1g, 105°C, 3hr)
Specific optical rotation +104° to +110° (20°C, 589nm) (10 mg/ml in dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR