Fosetyl-aluminium

Supplier: Thermo Scientific Chemicals
Danger

46728-1000 467280250
TS46728-1000EA 230 USD
TS46728-1000 TS467280250
Fosetyl-aluminium
Fosetyl-aluminium
Formula: C₆H₁₈AlO₉P₃
Melting Pt: > 300°C
Storage Temperature: Ambient
MDL Number: MFCD00144376
CAS Number: 39148-24-8
EINECS: 254-320-2
Merck Index: 13.4275

Order Now

Specification Test Results

Appearance White crystalline powder
Infrared spectrum Conforms
Titration ≥ 94%
Water ≤ 1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR