Ropivacaine hydrochloride

Supplier: Thermo Scientific Chemicals

46722-0010 46722-2500
TS46722-0010EA 778 USD
TS46722-0010 TS46722-2500
Ropivacaine hydrochloride
Ropivacaine hydrochloride
Formula: C₁₇H₂₆N₂O·HCl
Melting Pt: 149°C…151°C
Storage Temperature: Refrigerator
MDL Number: MFCD06671728
CAS Number: 98717-15-8
Merck Index: 13.8339

Order Now

Specification Test Results

Appearance White to beige powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Water ≤ 6.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR