Telaprevir

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1S,3aR,6aS)-2-((S)-2-((S)-2-Cyclohexyl-2-(pyrazine-2-carboxamido)acetamido)-3,3-dimethylbutanoyl)-N-((S)-1-(cyclopropylamino)-1,2-dioxohexan-3-yl)octahydrocyclopenta[c]pyrrole-1-carboxamide, (3S,3aS,6aR)-2-[(2S)-2-[[(2S)-2-cyclohexyl-2-(pyrazine-2-carbonylamino)acetyl]amino]-3,3-dimethylbutanoyl]-N-[(3S)-1-(cyclopropylamino)-1,2-dioxohexan-3-yl]-3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrole-3-carboxamide

46689-2500 466890010
TS46689-2500EA 265 USD
TS46689-2500 TS466890010
Telaprevir
Telaprevir
Formula: C₃₆H₅₃N₇O₆
MW: 679.85 g/mol
Melting Pt: 221°C…224°C
Storage Temperature: Freezer
MDL Number: MFCD11616089
CAS Number: 402957-28-2

Order Now

Specification Test Results

Appearance White to light yellow powder
Infrared spectrum Conforms
HPLC ≥ 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR