Triptolide

Supplier: Thermo Scientific Chemicals
Danger

46684-0250 466841000
TS46684-0250EA 370 USD
TS46684-0250 TS466841000
Triptolide
Triptolide
Formula: C₂₀H₂₄O₆
MW: 360.41 g/mol
Melting Pt: 226°C…227°C
Storage Temperature: Refrigerator
MDL Number: MFCD00210565
CAS Number: 38748-32-2

Order Now

Specification Test Results

Appearance White to almost white powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥ 97.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR