Gentiopicroside

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5R,6S)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-vinyl-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, Gentiopicrin

46683-2500 466830010
TS46683-2500EA 232 USD
TS46683-2500 TS466830010
Gentiopicroside
Gentiopicroside
Formula: C₁₆H₂₀O₉
MW: 356.33 g/mol
Melting Pt: 191°C
Storage Temperature: Refrigerator
MDL Number: MFCD00075700
CAS Number: 20831-76-9
EINECS: 244-070-2
Merck Index: 13.4408

Order Now

Specification Test Results

Appearance White to light yellow powder
Infrared spectrum Conforms
HPLC ≥ 98.0%
Water ≤ 2.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR