Idebenone

Supplier: Thermo Scientific Chemicals

46677-0010 466770050
TS46677-0010EA 84 USD
TS46677-0010 TS466770050
Idebenone
Idebenone
Formula: C₁₉H₃₀O₅
Melting Pt: 52°C…55°C
Storage Temperature: Ambient
MDL Number: MFCD00274552
CAS Number: 58186-27-9
Merck Index: 13.4913

Order Now

Specification Test Results

Appearance Yellow to orange powder
Infrared spectrum Conforms
Melting point 51.0°C to 55.0°C
HPLC ≥ 98.0%
Loss on drying ≤ 0.5%
Solubility (5% in methanol) Clear to slight yellow to orange solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR