Adenosine 5'-diphosphate disodium salt (ADP disodium salt) hydrate 98%

Supplier: Thermo Scientific Chemicals

16467-0010 16467-0050 16467-2500
TS16467-0010EA 104 USD
TS16467-0010 TS16467-0050 TS16467-2500
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) hydrate 98%
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate
Formula: C₁₀H₁₃N₅Na₂O₁₀P₂·xH₂O
MW: 471.167 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150927
CAS Number: 16178-48-6
EINECS: 240-314-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤10 % (K.F.)
Solubility (5 % w/v in water) Clear colorless
UV spectrophotometry molar absorptivity 15000 +/- 2%, pH 7
Ratio of Absorbances 0.15 to 0.17 (A280/A260)
Sodium (Na) 8 to 10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR