Isoprenaline hemisulfate

Supplier: Thermo Scientific Chemicals

Synonyms: 4-[1-Hydroxy-2-[(1-isopropyl)amino]ethyl]-1,2-benzenediol hemisulphate

46211-0050 46211-0250
TS46211-0050EA 53 USD
TS46211-0050 TS46211-0250
Isoprenaline hemisulfate
Isoprenaline hemisulphate
Formula: C₁₁H₁₇NO₃·¹/₂H₂O₄S
MW: 260.30 g/mol
Melting Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00013139
CAS Number: 299-95-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Water ≤7 %
HPLC ≥97.5 %
Identity passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR