Ganciclovir 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9-[[2-Hydroxy-1-(hydroxymethyl)ethoxy]methyl]guanine, 2'-Nor-2'-Deoxyguanosine, BW 759, 2-Amino-9-(((1,3-dihydroxypropan-2-yl)oxy)methyl)-1H-purin-6(9H)-one, RS-21592

46171-0010 46171-0050 46171-0250
TS46171-0010EA 42 USD
TS46171-0010 TS46171-0050 TS46171-0250
Ganciclovir 98%
Ganciclovir
Formula: C₉H₁₃N₅O₄
MW: 255.23 g/mol
Melting Pt: 251…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00870588
CAS Number: 82410-32-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤4 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR