Chlorogenic acid predominantly trans (from coffee seeds) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

10924-0050 10924-0010
TS10924-0050EA 440 USD
TS10924-0050 TS10924-0010
Chlorogenic acid predominantly trans (from coffee seeds) 98%
Chlorogenic acid
Formula: C₁₆H₁₈O₉
MW: 354.31 g/mol
Melting Pt: 205…209 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00003862
CAS Number: 327-97-9

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR