Acarbose 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Prandase, Acarbose Hydrate, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay

45908-0010 45908-0050
TS45908-0010EA 110 USD
TS45908-0010 TS45908-0050
Acarbose 95%
Acarbose
Formula: C₂₅H₄₃NO₁₈
MW: 645.61 g/mol
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00869592
CAS Number: 56180-94-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥94 %
Specific optical rotation +168° to +183° (20°C, 589 nm) (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR