Cytidine-5'-monophosphate (from yeast) 97%

Supplier: Thermo Scientific Chemicals

44949-0010 44949-0050
TS44949-0010EA 82 USD
TS44949-0010 TS44949-0050
Cytidine-5'-monophosphate (from yeast) 97%
Cytidine-5'-monophosphate
Formula: C₉H₁₄N₃O₈P
Melting Pt: 222 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006544
CAS Number: 63-37-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤3 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR