1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 98+%
Supplier: Thermo Scientific Chemicals
11201-0250
11201-1000
11201-5000
TS11201-0250EA
52
USD
TS11201-0250
TS11201-1000
TS11201-5000
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose 98+%
1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose
Formula:
C₁₂H₂₀O₆ Melting Pt: 107…110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005544 CAS Number: 582-52-5 |
Specification Test Results
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.2 % |
GC | ≥98.0 % |
Specific optical rotation | -10° to -12° (20°C, 589 nm) (c=5, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...