Nystatin 85+%
Supplier: Thermo Scientific Chemicals
45550-0010
45550-0050
45550-0250
TS45550-0010EA
31.02
USD
TS45550-0010
TS45550-0050
TS45550-0250
Nystatin 85+%
Nystatin
Formula:
C₄₇H₇₅NO₁₇ Melting Pt: >160 °C Storage Temperature: Freezer |
MDL Number:
MFCD25541651 CAS Number: 1400-61-9 |
Specification Test Results
Appearance | Yellow to light brown Powder |
Infrared spectrum | Conforms |
Water | ≤6 % |
HPLC | ≥85.0 % |
UV | at 305 nm A: ≥0.60 |
Activity | ≥4400 IU/mg (dried substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...