Guanosine 5'-triphosphate disodium salt hydrate 90% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: GTP disodium salt hydrate

22625-0010 22625-0500 22625-2500
TS22625-0010EA 632 USD
TS22625-0010 TS22625-0500 TS22625-2500
Guanosine 5'-triphosphate disodium salt hydrate 90% for biochemistry
Guanosine 5'-triphosphate disodium salt hydrate
Formula: C₁₀H₁₄N₅Na₂O₁₄P₃·H₂O
MW: 567.147 g/mol (anhy)
Melting Pt: 180 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD03410297
CAS Number: 56001-37-7
EINECS: 259-940-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤12 % (K.F.)
HPLC ≥88 %
UV spectrophotometry ≥88 % (lambda max. 252 nm, pH 7)
Ratio of Absorbances 0.63 to 0.69 (A280/A260)
Related substances ≤4 % (typically) Guanosine diphosphate (HPLC)
Sodium (Na) 7 to 9.5 % (flame photom.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR