Guanosine-5'-monophosphate disodium salt 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium 5'-guanylate, 5'-Guanosine monophosphate disodium salt, GMP disodium salt, 5'-GMP 2Na, 5'-Guanylic acid disodium salt, Guanosine-5'-Monophosphate

22660-0050 22660-0250
TS22660-0050EA 101 USD
TS22660-0050 TS22660-0250
Guanosine-5'-monophosphate disodium salt 97%
Guanosine-5'-monophosphate disodium salt
Formula: C₁₀H₁₂N₅Na₂O₈P
MW: 407.187 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00151238
CAS Number: 5550-12-9
EINECS: 226-914-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤25 % (120°C, 4 h)
TLC Homogeneous
UV spectrophotometry molar absorptivity 13700 +/- 4%, pH 7
Titration with HClO4 ≥96.0 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR