Guanosine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
41113-0250
41113-1000
41113-5000
TS41113-0250EA
70
USD
TS41113-0250
TS41113-1000
TS41113-5000
Guanosine 99%
Guanosine
Formula:
C₁₀H₁₃N₅O₅ MW: 283.24 g/mol Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010182 CAS Number: 118-00-3 EINECS: 204-227-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % |
Titration with HClO4 | ≥98.5 % (On dry substance) |
Specific optical rotation | -9.5° to -12.5° (20°C, 589 nm) (c=1, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...