N-Lauroylsarcosine sodium salt 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

43437-0010 43437-0250 43437-1000
TS43437-0010EA 490 USD
TS43437-0010 TS43437-0250 TS43437-1000
N-Lauroylsarcosine sodium salt 95%
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MW: 293.38 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
UN: 2811
ADR: 6.1 II
Merck Index: 12,04379
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to almost white Powder
Infrared spectrum Conforms
Water ≤7 %
Solubility (10 % in water) Almost clear colorless to light yellow
pH 7.0 to 9.0
Titration with HClO4 94 to 106 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR