5-Hydroxy-L-tryptophan 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, 5-Hydroxyl-L-tryptophan, (-)-5-Hydroxy-L-Tryptophane, L-5-Hydroxytryptophan, Oxitriptan, H-Trp(5-OH)-OH

14829-0010 14829-0050 14829-0250
TS14829-0010EA 41 USD
TS14829-0010 TS14829-0050 TS14829-0250
5-Hydroxy-L-tryptophan 99%
5-Hydroxy-L-tryptophan
Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064341
CAS Number: 4350-09-8
EINECS: 224-411-1
UN: 2811
ADR: 6.1,III
Merck Index: 12,04895

Order Now

Specification Test Results

Appearance White to light beige to light grey Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Heavy metals ≤10 ppm
Sulfated ash ≤0.3 %
HPLC ≥98.5 % (on dry substance)
Specific optical rotation -30.9° to -34.9° (20°C, 589 nm) (c=1, H2O) (on dry substance)
Titration with HClO4 98.5 to 101.5 % (On dry substance)
Arsenic (As) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR