Eriodictyol

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-[3,4-Dihydroxyphenyl]2,3-dihydro-5,7-dihydroxy-4H-1benzopyran-4-one), (S)-2-(3,4-Dihydroxyphenyl)-5,7-dihydroxychroman-4-one

46524-2500 465240010
TS46524-2500EA 179 USD
TS46524-2500 TS465240010
Eriodictyol
Eriodictyol
Formula: C₁₅H₁₂O₆
MW: 288.252 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00135890
CAS Number: 552-58-9

Order Now

Specification Test Results

Appearance White to light beige to light brown Powder or crystals
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR