Scutellarin

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3S,4S,5R,6S)-6-((5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid

46513-0010 465132500
TS46513-0010EA 522 USD
TS46513-0010 TS465132500
Scutellarin
Scutellarin
Formula: C₂₁H₁₈O₁₂
MW: 462.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01861503
CAS Number: 27740-01-8

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR