Indacaterol maleate

Supplier: Thermo Scientific Chemicals

46465-2500 464650010
TS46465-2500EA 185 USD
TS46465-2500 TS464650010
Indacaterol maleate
Indacaterol maleate
Formula: C₂₄H₂₈N₂O₃·C₄H₄O₄
MW: 508.57 g/mol
Melting Pt: 195…197 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD20526769
CAS Number: 753498-25-8

Order Now

Specification Test Results

Appearance Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR