Sitafloxacin sesquihydrate

Supplier: Thermo Scientific Chemicals
Warning

46437-0010 46437-2500
TS46437-0010EA 393 USD
TS46437-0010 TS46437-2500
Sitafloxacin sesquihydrate
Sitafloxacin sesquihydrate
Formula: C₁₉H₁₈ClF₂N₃O₃·1.5H₂O
MW: 436.84 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00903720
CAS Number: 163253-35-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying 5.5 % to 8.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR