Coenzyme B12

Supplier: Thermo Scientific Chemicals

46436-2500 464360010
TS46436-2500EA 121 USD
TS46436-2500 TS464360010
Coenzyme B12
Coenzyme B12
Formula: C₇₂H₁₀₀CoN₁₈O₁₇P
Storage Temperature: Freezer
MDL Number: MFCD00135609
CAS Number: 13870-90-1

Order Now

Specification Test Results

Appearance Red to brown Powder
Infrared spectrum Conforms
Water ≤12.0 %
Solubility (20 mg/ml in water) Clear light to dark red solution
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR