Acetamiprid

Supplier: Thermo Scientific Chemicals
Danger

46393-0010 46393-2500
TS46393-0010EA 410 USD
TS46393-0010 TS46393-2500
Acetamiprid
Acetamiprid
Formula: C₁₀H₁₁ClN₄
MW: 222.67 g/mol
Melting Pt: 100…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06201842
CAS Number: 135410-20-7
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 95 to 102 °C
Water ≤1 %
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR