Oxybutynin hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxybutynin chloride, 4-(Diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride

46350-0010 46350-0050 463500250
TS46350-0010EA 123 USD
TS46350-0010 TS46350-0050 TS463500250
Oxybutynin hydrochloride
Oxybutynin hydrochloride
Formula: C₂₂H₃₁NO₃·HCl
MW: 393.95 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00072150
CAS Number: 1508-65-2

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤3 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR