Divalproex sodium

Supplier: Thermo Scientific Chemicals
Danger

46349-0250 463491000
TS46349-0250EA 113 USD
TS46349-0250 TS463491000
Divalproex sodium
Divalproex sodium
Formula: C₁₆H₃₁NaO₄
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00879852
CAS Number: 76584-70-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
Water ≤1 %
HPLC ≥97.5 %
Sodium (Na) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR