Emamectin benzoate

Supplier: Thermo Scientific Chemicals
Danger

46305-0010 463050050
TS46305-0010EA 131 USD
TS46305-0010 TS463050050
Emamectin benzoate
Emamectin benzoate
Formula: C₄₉H₇₅NO₁₃·C₇H₆O₂
Melting Pt: 146…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027777
CAS Number: 155569-91-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow to light beige Powder or crystals
Infrared spectrum Conforms
Water ≤3 %
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR