Tylosin phosphate

Supplier: Thermo Scientific Chemicals
Danger

46304-0050 463040010
TS46304-0050EA 249 USD
TS46304-0050 TS463040010
Tylosin phosphate
Tylosin phosphate
Formula: C₄₆H₇₇NO₁₇·H₃PO₄
Storage Temperature: Refrigerator
MDL Number: MFCD06795692
CAS Number: 1405-53-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Solubility (1% in water) Clear colorless to light yellow
HPLC ≥94 % (sum of tylosins A+B+C+D)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR