Vitamin B12a hydrochloride

Supplier: Thermo Scientific Chemicals

Synonyms: Hydroxocobalamin hydrochloride, Hydroxocobalamin xhydrochloride

46208-0010 462080050
TS46208-0010EA 276 USD
TS46208-0010 TS462080050
Vitamin B12a hydrochloride
Vitamin B12a hydrochloride
Formula: C₆₂H₈₉CoN₁₃O₁₅P·HCl
MW: 1346.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00167463
CAS Number: 58288-50-9
New Product

Order Now

Specification Test Results

Appearance Dark red to dark purple Crystalline powder or crystals
Infrared spectrum Conforms
Assay ≥85 %
Loss on drying ≤12 %
HPLC ≥90 %
Identification Chlorides: passes test
UV Lambda max 525 nm
Absorbance scale 0.31 to 0.35 (A525/A351)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR